missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Violet 2B

Product Code. 11410143
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410143 50 g 50g
11420143 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410143 Supplier Thermo Scientific Chemicals Supplier No. A12159.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 8004-87-3 | C24H27N3·HCl

Methyl violet 2B is a biological stain that is commonly used at a pH between 0 and 1.6. It is certified for use in Flemming triple stain with iodine for chromosomes; Newton?s crystal violet-iodine technique for chromatin and nucleoli; Lieb?s amyloid stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl violet 2B is a biological stain that is commonly used at a pH between 0 and 1.6. It is certified for use in Flemming triple stain with iodine for chromosomes; Newton′s crystal violet-iodine technique for chromatin and nucleoli; Lieb′s amyloid stain.

Solubility
Soluble in water.

Notes
Transition interval: pH 0.0 (yellow) to pH 1.6 (blue)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8004-87-3
Molecular Formula C24H27N3·HCl
MDL Number MFCD00012568
Synonym Basic Violet 1; C.I. 42535

Specifications

Solubility Information Soluble in water.
Formula Weight 393.96
Odor Odorless
Quantity 50 g
Chemical Name or Material Methyl Violet 2B
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyl Violet 2B
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: C.I. Basic Violet

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.