missing translation for 'onlineSavingsMsg'
Learn More

Methyl vinyl sulfone, 95%, stabilized

Product Code. 10055773 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055773 50 g Glass bottle 50g
10311244 1 g Glass bottle 1g
10678211 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10055773 Supplier Thermo Scientific Acros Supplier No. 306730500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3680-02-2 | C3H6O2S | 106.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3680-02-2
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00007567
InChI Key WUIJTQZXUURFQU-UHFFFAOYSA-N
ChEBI CHEBI:46938
SMILES CS(=O)(=O)C=C

Specifications

Color Colorless to Yellow
Density 1.2100g/mL
Boiling Point 115.0°C to 117.0°C (19.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHSO2CH3
Refractive Index 1.4610 to 1.4630
Quantity 50 g
Specific Gravity 1.21
Synonym zebularine, pyrimidin-2-one ribonucleoside, pyrimidin-2-one beta-ribofuranoside, pyrimidin-2-one beta-d-ribofuranoside, 4-deoxyuridine, 1-beta-d-ribofuranosyl-2 1h-pyrimidinone, 1-beta-d-ribofuranosylpyrimidin-2 1h-one, unii-7a9y5sx0gy, 7a9y5sx0gy, 2 1h-pyrimidinone, 1-beta-d-ribofuranosyl
IUPAC Name methanesulfonylethene
Stabilizer 150ppm 4-tert.-butylpenol
PubChem CID 100016
Formula Weight 106.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl vinyl sulfone, Stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.