missing translation for 'onlineSavingsMsg'
Learn More

Methyl vinyl sulfone, 95%, stab. with 200ppm 4-tert-butylphenol

Product Code. 11381307
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11381307 5 g 5g
11371307 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11381307 Supplier Thermo Scientific Alfa Aesar Supplier No. L01519.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3680-02-2 | C3H6O2S | 106.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3680-02-2
Molecular Formula C3H6O2S
Molecular Weight (g/mol) 106.14
MDL Number MFCD00007567
InChI Key WUIJTQZXUURFQU-UHFFFAOYSA-N
Synonym methyl vinyl sulfone, ethene, methylsulfonyl, methyl vinyl sulphone, methylvinylsulfone, methanesulfonylethene, sulfone, methyl vinyl, vinyl methyl sulfone, methanesulfonyl-ethene, methylsulfonyl ethene, methyl vinyl sulfphone
PubChem CID 19354
IUPAC Name 1-methylsulfonylethene
SMILES CS(=O)(=O)C=C

Specifications

Density 1.212
Boiling Point 115°C to 117°C (19 mmHg)
Flash Point >109°C (228°F)
Refractive Index 1.463
Quantity 5 g
UN Number UN2810
Beilstein 878198
Formula Weight 106.15
Percent Purity 95%
Chemical Name or Material Methyl vinyl sulfone, stabilized with 200ppm 4-tert-butylphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.