missing translation for 'onlineSavingsMsg'
Learn More

Methyl vinyl ketone, tech. 90%, stab.

Product Code. 11451890
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451890 100 mL 100mL
11441890 25 mL 25mL
11461890 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11451890 Supplier Thermo Scientific Alfa Aesar Supplier No. A11910.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 78-94-4 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-94-4
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00008777
InChI Key FUSUHKVFWTUUBE-UHFFFAOYSA-N
Synonym methyl vinyl ketone, 3-buten-2-one, butenone, methylvinylketon, vinyl methyl ketone, methylene acetone, 2-butenone, acetyl ethylene, 3-butene-2-one, methyl ethenyl ketone
PubChem CID 6570
ChEBI CHEBI:48058
IUPAC Name but-3-en-2-one
SMILES CC(=O)C=C

Specifications

Melting Point °C to 7°C
Density 0.856
Boiling Point 32°C to 34°C (60 mm)
Flash Point -6°C (21°F)
Assay Percent Range 90%, Stabilized with 0.5% Hydroquinone
Refractive Index 1.412
Quantity 100 mL
UN Number UN1251
Beilstein 506021
Merck Index 14,6134
Formula Weight 70.09
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Methyl Vinyl Ketone, Stabilized
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.