missing translation for 'onlineSavingsMsg'
Learn More

Methyl undecanoate, 99%

Product Code. 11362296
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362296 25 g 25g
11352296 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362296 Supplier Thermo Scientific Alfa Aesar Supplier No. L05939.14
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1731-86-8 | C12H24O2 | 200.322 g/mol

Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl undecanoate is used as plasticizer in polymers. It is used as fragrance agent in aroma and flavor Chemicals.

Solubility
Soluble in ethanol, and ether. Insoluble in water.

Notes
Store at -20°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1731-86-8
Molecular Formula C12H24O2
Molecular Weight (g/mol) 200.322
MDL Number MFCD00008957
InChI Key XPQPWPZFBULGKT-UHFFFAOYSA-N
Synonym undecanoic acid methyl ester, undecanoic acid, methyl ester, methyl n-undecanoate, unii-k67yz97w6c, undecanoic acid methylester, n-undecanoic acid methyl ester, methyl undecanoate, acmc-209e5z, undecanoic acid, methylester, 3-02-00-00858 beilstein handbook reference
PubChem CID 15607
ChEBI CHEBI:87527
IUPAC Name methyl undecanoate
SMILES CCCCCCCCCCC(=O)OC

Specifications

Melting Point -10°C
Density 0.871
Boiling Point 246°C to 248°C
Flash Point 109°C (228°F)
Refractive Index 1.429
Quantity 25 g
Beilstein 1762129
Solubility Information Soluble in ethanol,and ether. Insoluble in water.
Percent Purity 99%
Chemical Name or Material Methyl undecanoate
compliance-icons
Product Identifier
  • Methyl undecanoate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.