missing translation for 'onlineSavingsMsg'
Learn More

Methyl (triphenylphosphoranylidene)acetate, 98%

Product Code. 10664842 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10664842 25 g Glass bottle 25g
10702791 100 g Glass bottle 100g
11995801 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10664842 Supplier Thermo Scientific Acros Supplier No. 169560250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2605-67-6 | C21H19O2P | 334.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2605-67-6
Molecular Formula C21H19O2P
Molecular Weight (g/mol) 334.36
MDL Number MFCD00008455
InChI Key NTNUDYROPUKXNA-UHFFFAOYSA-N
Synonym methyl triphenylphosphoranylidene acetate, carbomethoxymethylene triphenylphosphorane, methyl 2-triphenylphosphoranylidene acetate, acetic acid, triphenylphosphoranylidene-, methyl ester, methyl triphenylphosphoranylideneacetate, triphenylphosphoranylidene acetic acid methyl ester, triphenyl carbomethoxymethylene phosphorane, methoxycarbonylmethylene triphenylphosphorane, acetic acid, 2-triphenylphosphoranylidene-, methyl ester
PubChem CID 17453
IUPAC Name methyl 2-(triphenyl-$l^{5}-phosphanylidene)acetate
SMILES COC(=O)C=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164.0°C to 169.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 2% max. triphenylphosphine+ triphenylphosphine oxide (HPLC)
Packaging Glass bottle
Linear Formula (C6H5)3P=CHCO2CH3
Quantity 25 g
Fieser 01,112; 01,1081; 08,456; 09,426; 10,363; 13,277
Solubility Information Solubility in water: insoluble
Formula Weight 334.34
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl (triphenylphosphoranylidene)acetate
Show More Show Less
compliance-icons
Product Identifier
  • Methyl (triphenylphosphoranylidene)acetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.