missing translation for 'onlineSavingsMsg'
Learn More

Methyl (trimethylsilyl)propiolate, 98%

Product Code. 11480778
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480778 5 g 5g
11470778 1 g 1g
11490778 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480778 Supplier Thermo Scientific Alfa Aesar Supplier No. B25245.06
 Restricted Item
Estimated Shipment: 02-12-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 42201-71-8 | C7H12O2Si | 156.256 g/mol

Methyl (trimethylsilyl)propiolate is used as the intermediates of OLED, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl (trimethylsilyl)propiolate is used as the intermediates of OLED, pharmaceuticals.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 42201-71-8
Molecular Formula C7H12O2Si
Molecular Weight (g/mol) 156.256
MDL Number MFCD04039783
InChI Key XXGHAMBKHIDZDC-UHFFFAOYSA-N
Synonym methyl trimethylsilyl propiolate, methyl 3-trimethylsilylpropynoate, methyl 3-trimethylsilyl propiolate, methyl 3-trimethylsilyl-2-propynoate, acmc-20akhx, methyl 3-trimethylsilylpropiolate, methyl 3-trimethylsilyl propynoate, trimethylsilylpropiolic acid methyl ester, trimethylsilyl propiolic acid methyl ester, methyl 3-trimethylsilyl-2-propynoate #
PubChem CID 560731
IUPAC Name methyl 3-trimethylsilylprop-2-ynoate
SMILES COC(=O)C#C[Si](C)(C)C

Tekniske data

Boiling Point 71°C to 75°C (11mmHg)
Quantity 5 g
Refractive Index 1.44
Linear Formula (CH3)3SiC≡CCO2CH3
UN Number UN3272
Formula Weight 156.25
Percent Purity 98%
Chemical Name or Material Methyl (trimethylsilyl)propiolate

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.