missing translation for 'onlineSavingsMsg'
Learn More

Methyl trimethylacetate, 99%

Product Code. 11444814
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444814 500 g 500g
11434814 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444814 Supplier Thermo Scientific Alfa Aesar Supplier No. A13764.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 598-98-1 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-98-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008843
InChI Key CNMFHDIDIMZHKY-UHFFFAOYSA-N
Synonym methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3
PubChem CID 69027
IUPAC Name methyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC

Specifications

Melting Point <-70°C
Density 0.875
Boiling Point 100°C to 101°C
Flash Point 6°C (42°F)
Refractive Index 1.39
Quantity 500 g
UN Number UN3272
Beilstein 1744141
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material Methyl trimethylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.