missing translation for 'onlineSavingsMsg'
Learn More

Methyl trifluoropyruvate, 97%

Product Code. 11497194
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497194 25 g 25g
11477194 1 g 1g
11487194 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497194 Supplier Thermo Scientific Alfa Aesar Supplier No. B21589.14
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13089-11-7 | C4H3F3O3 | 156.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13089-11-7
Molecular Formula C4H3F3O3
Molecular Weight (g/mol) 156.06
MDL Number MFCD00114936
InChI Key XGLLQDIWQRQROJ-UHFFFAOYSA-N
Synonym methyl trifluoropyruvate, methyl 3,3,3-trifluoropyruvate, methyltrifluoropyruvate, trifluoropyruvic acid methyl ester, 3,3,3-trifluoropyruvic acid methyl ester, propanoic acid, 3,3,3-trifluoro-2-oxo-, methyl ester, pubchem2012, rarechem al bf 1300, acmc-1bp0j
PubChem CID 2734931
IUPAC Name methyl 3,3,3-trifluoro-2-oxopropanoate
SMILES COC(=O)C(=O)C(F)(F)F

Specifications

Density 1.52
Boiling Point 84°C to 86°C
Flash Point 26°C (79°F)
Refractive Index 1.332
Quantity 25 g
UN Number UN3272
Beilstein 1863490
Formula Weight 156.06
Percent Purity 97%
Chemical Name or Material Methyl trifluoropyruvate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.