missing translation for 'onlineSavingsMsg'
Learn More

Methyl trifluoromethanesulfonate, 97%

Product Code. 15448328
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448328 100 g 100g
15438328 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448328 Supplier Thermo Scientific Alfa Aesar Supplier No. H61758.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 333-27-7 | C2H3F3O3S | 164.10 g/mol

Methyl trifluoromethanesulfonate is used as a powerful methylating reagent in chemistry. Further, it is used in the conversion of amines to methyl ammonium triflates. In addition, it is also used in Suzuki reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl trifluoromethanesulfonate is used as a powerful methylating reagent in chemistry. Further, it is used in the conversion of amines to methyl ammonium triflates. In addition, it is also used in Suzuki reaction.

Notes
Air and moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong bases, strong acids and strong reducing agents. Hydrolyzes with water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-27-7
Molecular Formula C2H3F3O3S
Molecular Weight (g/mol) 164.10
MDL Number MFCD00000409
InChI Key OIRDBPQYVWXNSJ-UHFFFAOYSA-N
Synonym methyl triflate, methyl trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, methyl ester, triflate ester, methyl trifluoromethane sulfonate, trifluoromethanesulfonic acid methyl ester, unii-7b25z22epv, ccris 1158, methyltrifluoromethanesulfonate, methanesulfonic acid, 1,1,1-trifluoro-, methyl ester
PubChem CID 9526
IUPAC Name methyl trifluoromethanesulfonate
SMILES COS(=O)(=O)C(F)(F)F

Specifications

Density 1.5
Boiling Point 99°C
Flash Point 38°C (100°F)
Linear Formula CF3SO3CH3
Refractive Index 1.326
Quantity 100 g
UN Number UN2920
Sensitivity Air sensitive
Formula Weight 164.1
Percent Purity 98%
Chemical Name or Material Methyl trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.