missing translation for 'onlineSavingsMsg'
Learn More

Methyl tridecanoate, 97%

Product Code. 10595100 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 10595100

Brand: Thermo Scientific Acros 438200050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1731-88-0 | C14H28O2 | 228.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1731-88-0
Molecular Formula C14H28O2
Molecular Weight (g/mol) 228.38
InChI Key JNDDPBOKWCBQSM-UHFFFAOYSA-N
Synonym tridecanoic acid, methyl ester, tridecanoic acid methyl ester, methyl n-tridecanoate, unii-o2h463ring, o2h463ring, tridecanoic acid,methyl ester, n-tridecanoic acid methyl ester, methyl tridecylate, methyltridecanoate, tridecanoic acid methyl
PubChem CID 15608
IUPAC Name methyl tridecanoate
SMILES CCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 7°C
Color Colorless
Density 0.8600g/mL
Boiling Point 130°C to 132°C (4.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.86
Formula Weight 228.38
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl tridecanoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.