missing translation for 'onlineSavingsMsg'
Learn More

Methyl tricosanoate, 99+%

Product Code. 10666460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10666460 1 g Glass bottle 1g
10461810 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10666460 Supplier Thermo Scientific Acros Supplier No. 428450010
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 2433-97-8 | C24H48O2 | 368.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2433-97-8
Molecular Formula C24H48O2
Molecular Weight (g/mol) 368.65
MDL Number MFCD00009349
InChI Key VORKGRIRMPBCCZ-UHFFFAOYSA-N
Synonym tricosanoic acid methyl ester, tricosanoic acid, methyl ester, unii-4v3pob2675, tricosanoic acid,methyl ester, tricosanoic acid methyl ester, n, methyl tricosanoate gc, methyl tricosanoate, analytical standard, ffdf5aad-7f50-410b-ae3f-fafbe527e4bc, tricosanoic acid methyl ester 10 microg/ml in cyclohexane, methyl tricosanoate, united states pharmacopeia usp reference standard
PubChem CID 75519
IUPAC Name methyl tricosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 53°C
Flash Point >113°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)21CO2CH3
Quantity 1 g
Formula Weight 368.64
Percent Purity 99+%
Chemical Name or Material Methyl tricosanoate
compliance-icons
Product Identifier
  • Methyl tricosanoate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.