missing translation for 'onlineSavingsMsg'
Learn More

Methyl trans-2-pentenoate, 95%

Product Code. 10662192 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Plastic bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10662192 5 g Plastic bottle 5g
10111091 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10662192 Supplier Thermo Scientific Supplier No. 329720050
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Læg i indkøbskurv
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 15790-88-2 | C6H10O2 | 114.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 15790-88-2
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00137611
InChI Key MBAHGFJTIVZLFB-SNAWJCMRSA-N
Synonym methyl 2-pentenoate, methyl trans-2-pentenoate, methyl 2e-pent-2-enoate, 2-pentenoic acid, methyl ester, 2-pentenoic acid, methyl ester, e, methyl 3-ethylacrylate, methyl 2-transpentenoate, methyl 2-trans-pentenoate, methyl e-2-pentenoate, trans-methyl pent-2-enoate
PubChem CID 5364718
SMILES CC\C=C\C(=O)OC

Tekniske data

Color Colorless
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic bottle
Refractive Index 1.4300 to 1.4320
Quantity 5 g
IUPAC Name methyl (2E)-pent-2-enoate
Formula Weight 114
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl trans-2-pentenoate, 95%
Vis mere Vis mindre
compliance-icons compliance-icons
Produktidentifikator
  • Methyl trans-2-pentenoate
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.