missing translation for 'onlineSavingsMsg'
Learn More

Methyl thiophene-3-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15448038
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448038 5 g 5g
15438038 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448038 Supplier Thermo Scientific Alfa Aesar Supplier No. H61488.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 22913-26-4 | Molecular Formula: C6H6O2S | Molecular Weight: 142.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22913-26-4
Molecular Formula C6H6O2S
Molecular Weight (g/mol) 142.17
MDL Number MFCD06203810
InChI Key ZTRAEMILTFNZSM-UHFFFAOYSA-N
Synonym methyl 3-thiophenecarboxylate, 3-thiophenecarboxylic acid, methyl ester, thiophene-3-carboxylic acid methyl ester, 3-thiophenecarboxylicacid, methyl ester, methyl thiophen-3-carboxylate, methyl-3-thenoate, methyl 3-thenoate, 3-methoxycarbonylthiophene, 3-methoxy carbonyl-thiophene, methyl 3-thiophenecarboxylate #
PubChem CID 574151
IUPAC Name methyl thiophene-3-carboxylate
SMILES COC(=O)C1=CSC=C1

Specifications

Density 1.174
Boiling Point 98°C (10 mmHg)
Flash Point 77°C (171°F)
Refractive Index 1.538
Quantity 5 g
Formula Weight 284.35
Percent Purity 97%
Chemical Name or Material Methyl thiophene-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.