missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Thiophene-2-carboxylate, Mikromol™
Description
Methyl Thiophene-2-carboxylate
Specifications
Specifications
| Chemical Name or Material | Methyl Thiophene-2-carboxylate |
| CAS | 5380-42-7 |
| Analyte or Component Names | Methyl Thiophene-2-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H6 O2 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl Thiophene-2-carboxylate, 2-Thiophenecarboxylic acid methyl ester, 2-(Carbomethoxy)thiophene, 2-(Methoxycarbonyl)thiophene, Methyl 2-thienylcarboxylate, Methyl 2-thiophenecarboxylate, NSC 19879 |
| InChI Formula | InChI=1S/C6H6O2S/c1-8-6(7)5-3-2-4-9-5/h2-4H,1H3 |
| SMILES | COC(=O)c1cccs1 |
| IUPAC Name | methyl thiophene-2-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?