missing translation for 'onlineSavingsMsg'
Learn More

Methyl-Tert-Butyl Ether, Extra Pure, SLR, Fisher Chemical™

Product Code. 10428990
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber glass bottle
HDPE Plastic Bottle
Metal can
Plastic Coated Glass bottle

C5H12O, CAS Number-1634-04-4, methyl t-butyl ether, 2-methyl-2-methoxypropane, methyl tert-butyl ether, propane, 2-methoxy-2-methyl, methyl-t-butyl ether, mtbe, methyl tertiary-butyl ether, methyl-tert-butyl ether, tert-butyl methyl ether, t-butyl methyl ether, 5L, 54 deg.C, CHEBI:27642, Colorless

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1634-04-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number 8812
InChI Key BZLVMXJERCGZMT-UHFFFAOYSA-N
Synonym tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane
PubChem CID 15413
ChEBI CHEBI:27642
IUPAC Name 2-methoxy-2-methylpropane
SMILES CC(C)(C)OC

Specifications

Melting Point -110°C
Color Colorless
Boiling Point 54°C
Packaging Metal can
Quantity 25 L
Formula Weight 88.15g/mol
Vapor Pressure 268mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • tert-Butyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'