missing translation for 'onlineSavingsMsg'
Learn More

Methyl Syringate, TRC

Product Code. 30007911
Change view
Click to view available options
Quantity:
100 mg
25 g
100 g
Unit Size:
100g
100mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007911 100 g 100g
30006594 25 g 25g
30011176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007911 Supplier Toronto Research Chemicals Supplier No. TRCM266325100G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 884-35-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
Synonym 4-Hydroxy-3,5-dimethoxy-benzoic Acid Methyl Ester, Syringic Acid Methyl Ester, 4-Hydroxy-3,5-dimethoxybenzoic Acid Methyl Ester, DeniLite II Assist, Methyl 3,5-Dimethoxy-4-hydroxybenzoate, Methyl 4-Hydroxy-3,5-dimethoxybenzoate, Methyl Syringate, NSC 16946, NSC 611398, Novoprime F 258
IUPAC Name methyl 4-hydroxy-3,5-dimethoxybenzoate
SMILES O=C(OC)C1=CC(OC)=C(O)C(OC)=C1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H12O5/c1-13-7-4-6(10(12)15-3)5-8(14-2)9(7)11/h4-5,11H,1-3H3
Formula Weight 212.0685
Chemical Name or Material Methyl Syringate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.