Learn More
Methyl sulfoxide, 99.8+%, for HPLC, ACROS Organics™
Brand: Acros Organics 397600025
Code : 29
Additional Details : CAS Number : 67-68-5 Weight : 2.80000kg
Quantity | 2.5L |
---|
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
methylsulfinylmethane |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 | |
CS(=O)C |
Specifications
Methyl sulfoxide | |
87°C | |
1.4770 to 1.4800 | |
1.987 mPa.s (25°C) | |
18.4°C | |
For HPLC | |
99.8% min. (GC) | |
(CH3)2SO | |
15,3285 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,benzene,acetone,chloroform,,diethyl ether and aromatic hydrocarbons | |
CS(=O)C | |
78.13 | |
CHEBI:28262 | |
99.8+% |
1.1000g/mL | |
Glass bottle | |
1.1 | |
189.0°C | |
2.5L | |
67-68-5 | |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methylsulfinylmethane | |
679 | |
78.13 | |
HPLC |