missing translation for 'onlineSavingsMsg'
Learn More

Methyl sulfoxide-d6,

Product Code. 16338206 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16338206 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16338206 Supplier Thermo Scientific Acros Supplier No. 464071000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2D6OS | 84.18 g/

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Density 1.1900 g/mL
Flash Point 88°C
Specific Gravity 1.1900
Viscosity 2.4 mPa.s (20°C)
Quantity 100 mL
Melting Point 20.2°C
Boiling Point 190.0°C
CAS 2206-27-1
Molecular Formula C2D6OS
Solubility Information Solubility aq. soln.: soluble.
Molecular Weight (g/mol) 84.18
Grade NMR
Chemical Name or Material Methyl sulfoxide-d6
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.