missing translation for 'onlineSavingsMsg'
Learn More

Methyl Salicylate, MedChemExpress

Product Code. 16435467
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16435467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16435467 Supplier Medchem Express Supplier No. HYY0189
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Methyl Salicylate (Wintergreen oil) is a topical analgesic and anti-inflammatory agent. Also used as a pesticide, a denaturant, a fragrance ingredient, and a flavoring agent in food and tobacco products. A systemic acquired resistance (SAR) signal in tobacco. A topical nonsteroidal anti-inflammatory drug (NSAID). Methyl salicylate lactoside is a COX inhibitor.

Methyl Salicylate (Wintergreen oil) is a topical analgesic and anti-inflammatory agent. Also used as a pesticide, a denaturant, a fragrance ingredient, and a flavoring agent in food and tobacco products. A systemic acquired resistance (SAR) signal in tobacco. A topical nonsteroidal anti-inflammatory drug (NSAID). Methyl salicylate lactoside is a COX inhibitor.

Specifications

Chemical Name or Material Methyl Salicylate
CAS 119-36-8
Color Colorless
Purity Grade Notes Research
Molecular Formula C8H8O3
Quantity 50 mg
Synonym Wintergreen oil
Solubility Information Ethanol : 25 mg/mL (164.31 mM; Need ultrasonic)
SMILES O=C(OC)C1=CC=CC=C1O
Molecular Weight (g/mol) 152.15
Formula Weight 152.15
Grade Research
Physical Form Oil
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.