missing translation for 'onlineSavingsMsg'
Learn More

Methyl salicylate, 99%, Thermo Scientific Chemicals

Product Code. 10687744
Click to view available options
Quantity:
5 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-36-8
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00002214
InChI Key OSWPMRLSEDHDFF-UHFFFAOYSA-N
Synonym methyl salicylate, wintergreen oil, gaultheria oil, betula oil, teaberry oil, sweet birch oil, oil of wintergreen, analgit, spicewood oil, 2-hydroxybenzoic acid methyl ester
PubChem CID 4133
ChEBI CHEBI:31832
IUPAC Name methyl 2-hydroxybenzoate
SMILES COC(=O)C1=CC=CC=C1O

Specifications

Melting Point -8°C to 7°C
Color Colorless to Yellow
Density 1.1825g/mL
Boiling Point 222.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CO2CH3
Refractive Index 1.5350 to 1.5380
Quantity 2.5 L
Beilstein 10,7
Merck Index 15,6193
Specific Gravity 1.1825
Solubility Information Solubility in water: 0.07 % (20°C). Other solubilities: soluble in glacial acetic acid, soluble in ether and chloroform, in ethanol 80 vol. %:1:2 v/v (20°C), diethyl phthalate:1:1 v/v (20°C), 1,2-propanediol:1/7 v/v (20°C)
Formula Weight 152.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl salicylate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'