missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(+)-3-hydroxybutyrate, 99%, Thermo Scientific™

Product Code. 10387701 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10387701 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10387701 Supplier Thermo Scientific Acros Supplier No. 343860010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

CAS 53562-86-0
Color Colorless
Density 1.0550g/mL
Boiling Point 63°C (10.0 mmHg)
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H10O3
Refractive Index 1.4190 to 1.422
MDL Number MFCD00064461
Quantity 1 g
Specific Gravity 1.055
Specific Rotation Condition +21.5° (20°C c=neat)
Specific Rotation + 21.5
Synonym methyl s-+-3-hydroxybutyrate, s-methyl 3-hydroxybutanoate, methyl s-3-hydroxybutyrate, s-3-hydroxybutanoic acid methyl ester, unii-g10281a5ah, methyl 3s-3-hydroxybutanoate, butanoic acid, 3-hydroxy-, methyl ester, 3s, methyl-s-3-hydroxybutyrate, s-+-3-hydroxy-n-butyric acid methyl ester, s-3-hydroxybutyric acid methyl ester
InChI Key LDLDJEAVRNAEBW-UHFFFAOYNA-N
SMILES COC(=O)CC(C)O
IUPAC Name methyl (3S)-3-hydroxybutanoate
Molecular Weight (g/mol) 118.13
PubChem CID 6950307
Formula Weight 118.13
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl (S)-(+)-3-hydroxybutyrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.