missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl (S)-3-hydroxy-3-phenylpropanoate, 99%, Thermo Scientific™
Shop All Thermo Scientific Chemicals ProductsSpecifications
Specifications
| CAS | 36615-45-9 |
| Color | Colorless |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Molecular Formula | C10H12O3 |
| Refractive Index | 1.5180 to 1.52 |
| MDL Number | MFCD00221509 |
| Quantity | 1 g |
| Specific Rotation Condition | − 44.20 (21.00°C c=3,1,2−dichloroeth) |
| Show More |
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?