missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methyl (S)-(+)-3-hydroxy-2-methylpropionate, 99%

Brand:  Alfa Aesar™ 044483.04

Code : NEW

Additional Details : CAS Number : 80657-57-4 Weight : 0.00200kg

Quantity 2g

 View more versions of this product

Product Code. 11420830

  • £70.60 / 2g


Estimated Shipment date
from Supplier 12-03-2018
Add to basket

Specifications

Chemical Identifiers

Methyl (S)- (+)-3-hydroxy-2-methylpropionate
C5H10O3
MFCD00064520
(S)-3-Hydroxyisobutyric acid methyl ester; (S)-3-Hydroxy-2-methylpropionic acid methyl ester
80657-57-4
118.13
1720830

Specifications

Methyl (S)- (+)-3-hydroxy-2-methylpropionate
1.07g/mL
Liquid
C5H10O3
MFCD00064520
(S)-3-Hydroxyisobutyric acid methyl ester; (S)-3-Hydroxy-2-methylpropionic acid methyl ester
74°C (10mmHg)
81°C (178°F)
80657-57-4
1.4250
99%
118.13
1720830
Colorless
2g
+27° (c=4 in methanol)
Videos
SDS