missing translation for 'onlineSavingsMsg'
Learn More

Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate, 97%

Product Code. 15489058
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15489058

Brand: Thermo Scientific Alfa Aesar H62543.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4931-66-2 | C17H14F3N3S2 | 381.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4931-66-2
Molecular Formula C17H14F3N3S2
Molecular Weight (g/mol) 381.44
MDL Number MFCD00080931
InChI Key JLGLYOGXDCENSE-UHFFFAOYSA-N
Synonym methyl l-pyroglutamate, methyl s-+-2-pyrrolidone-5-carboxylate, l-pyroglutamic acid methyl ester, methyl pyroglutamate, methyl 5-oxo-l-prolinate, s-methyl 5-oxopyrrolidine-2-carboxylate, proline, 5-oxo-, methyl ester, methyl 2s-5-oxopyrrolidine-2-carboxylate, h-pyr-ome, s-+-methyl 2-pyrrolidone-5-carboxylate
PubChem CID 78646
IUPAC Name methyl (2S)-5-oxopyrrolidine-2-carboxylate
SMILES CN=C(NC1=CC=CC(=C1)C(F)(F)F)SCC1=CC=C(C=C1)N=C=S

Specifications

Color Yellow
Density 1.226 g/mL
Boiling Point 90°C (0.3 mmHg)
Flash Point 113°C (235°F)
Refractive Index 1.486
Quantity 5 g
Formula Weight 143.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.