missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red

Product Code. 11412688
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412688 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11412688 Supplier Thermo Scientific Alfa Aesar Supplier No. A16690.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 493-52-7 | C15H15N3O2 | 269.30 g/mol

Bacterial strains isolated from dye-contaminated sludge decolorized a toxic azo dye, namely, methyl red (MR). Methyl red is used in the methyl red test (MR test), used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bacterial strains isolated from dye-contaminated sludge decolorized a toxic azo dye, namely, methyl red (MR). Methyl red is used in the methyl red test (MR test), used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose

Solubility
Insoluble in water. Soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 179°C to 182°C
CAS 493-52-7
Molecular Formula C15H15N3O2
MDL Number MFCD00002425
Beilstein 750102
Merck Index 14,6119
Synonym C.I. 13020
Solubility Information Insoluble aq. soln.. Soluble in ethanol.
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=CC=C1C(O)=O
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
Molecular Weight (g/mol) 269.30
PubChem CID 10303
Formula Weight 269.3
Quantity 25 g
Chemical Name or Material Methyl Red
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.