missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Sodium Salt, TRC

Product Code. 30476266
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30476266 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30476266 Supplier Toronto Research Chemicals Supplier No. TRCM33285325G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Red Sodium Salt
CAS 845-10-3
Molecular Formula C15H15N3NaO2
Quantity 25 g
Synonym C.I. Acid Red 2, Sodium Salt, Methyl Red Sodium Salt, o-Methyl Red Sodium Salt, 2-[[4-(Dimethylamino)phenyl]azo]-benzoic Acid Sodium Salt
InChI Formula InChI=1S/C15H15N3O2.Na/c1-18(2)12-9-7-11(8-10-12)16-17-14-6-4-3-5-13(14)15(19)20;/h3-10H,1-2H3,(H,19,20);/q;+1/p-1
SMILES [Na+].CN(C)c1ccc(cc1)N=Nc2ccccc2C(=O)[O-]
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
Molecular Weight (g/mol) 292.29
Formula Weight 291.0984
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.