missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red, sodium salt, ACS reagent

Product Code. 10509983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10509983 100 g Glass bottle 100g
10065700 10 g Glass bottle 10g
10371491 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10509983 Supplier Thermo Scientific Chemicals Supplier No. 414841000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 845-10-3 | C15H14N3NaO2 | 291.286 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
MDL Number MFCD00002426
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
Synonym 2-[4-(Dimethylamino)phenylazo]benzoic acid, sodium salt, C.I. 13020, MR
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

Clarity (Aqueous) Passes Test
Linear Formula (CH3)2NC6H4N=NC6H4CO2Na
Beilstein 16, 329
Merck Index 15, 6192
Infrared Spectrum Authentic
Formula Weight 291.27
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval From pH 4.2 (pink) to pH 6.2 (yellow)
Color Index No. 13020
Color Orange/Red to Yellow
Quantity 100 g
Physical Form Liquid
Chemical Name or Material Methyl Red, sodium salt, Pure
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.