missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Red Mixed Solution According to European Pharmacopoeia (EP), Reagecon
Shop All Reagecon Products

Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Description
- European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
- High quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
- For manual titrations; indicator titrant or analyte change color and this color change is by far the most important method of end point detection, in such titrations.
- A high-quality indicator solution such as Reagecon's facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications
Specifications
| Packaging | Amber Glass Winchester Bottle |
| Quantity | 100 mL |
| Physical Form | Liquid |
Safety and Handling
Product Identifier
- METHYL RED MIXED SOLUTION
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
- MIXTURE LIST-Contains : ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction