missing translation for 'onlineSavingsMsg'
Learn More

Methyl Red Indicator, Reag. Ph. Eur., Honeywell Fluka™

Product Code. p-7120583 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15633830

Brand: Honeywell-Fluka 3265425G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reag. Ph. Eur., acid-base indicator

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym Acid Red 2
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O

Specifications

Melting Point 179°C to 182°C
Linear Formula (CH3)2NC6H4N=NC6H4CO2H
UN Number NONH for all modes of transport
Beilstein 750102
Solubility Information Slightly soluble in water
Formula Weight 269.3g/mol
Packaging Glass Bottle
Suitability Complies for sensitivity
Visual Transition Interval 4.4 to 6.0
Density 0.989g/cm3
Color Violet
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl Red, Reag. Ph. Eur.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.