missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red hydrochloride, ACS reagent

Product Code. 10206011 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10206011 25 g Glass bottle 25g
10479091 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10206011 Supplier Thermo Scientific Chemicals Supplier No. 339320250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 63451-28-5 | C15H16ClN3O2 | 305.762 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63451-28-5
Molecular Formula C15H16ClN3O2
Molecular Weight (g/mol) 305.762
MDL Number MFCD00012614
InChI Key ICTWKXAVJAHRMK-UHFFFAOYSA-N
Synonym Acid Red 2, 2-[4-(Dimethylamino)phenylazo]benzoic acid hydrochloride, C.I. 13020
PubChem CID 113227
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O.Cl

Specifications

Clarity Passes Test
Linear Formula (CH3)2NC6H4NNC6H4COOH·HCl
Beilstein 16, 329
Merck Index 15, 6192
Infrared Spectrum Authentic
Formula Weight 305.76
Grade ACS Reagent
Packaging Glass bottle
Visual Transition Interval from pH 4.2 (pink) to pH 6.2 (yellow)
Color Black to Blue
Quantity 25 g
Physical Form Powder
Chemical Name or Material Methyl Red hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.