missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(-)-mandelate, 99%

Product Code. 11328945
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328945 1 g 1g
11338945 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11328945 Supplier Thermo Scientific Alfa Aesar Supplier No. L14065.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 20698-91-3 | C9H10O3 | 166.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20698-91-3
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00064247
InChI Key ITATYELQCJRCCK-MRVPVSSYSA-N
Synonym methyl r---mandelate, r---methyl mandelate, r-methyl 2-hydroxy-2-phenylacetate, d---mandelic acid methyl ester, methyl 2r-2-hydroxy-2-phenylacetate, r-methyl mandelate, methyl d---mandelate, r-mandelic acid methyl ester, r---mandelic acid methyl ester, methyl hydroxy phenyl acetate #
PubChem CID 2724623
IUPAC Name methyl (2R)-2-hydroxy-2-phenylacetate
SMILES COC(=O)C(C1=CC=CC=C1)O

Specifications

Melting Point 56°C to 58°C
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 3589447
Optical Rotation −144° (c=1 in Methanol)
Formula Weight 166.18
Percent Purity 99%
Chemical Name or Material Methyl (R)-(-)-mandelate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.