missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(+)-lactate, 98%

Product Code. 10590934 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10590934 25 g Glass bottle 25g
10005712 1 g Glass bottle 1g
10219410 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10590934 Supplier Thermo Scientific Acros Supplier No. 414490250
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 17392-83-5 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17392-83-5
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD00004517
InChI Key LPEKGGXMPWTOCB-UHFFFAOYNA-N
Synonym methyl r-+-lactate, methyl d-lactate, r-methyl 2-hydroxypropanoate, methyl r-lactate, +-methyl d-lactate, methyl 2r-2-hydroxypropanoate, d-lactic acid methyl ester, methyl lactate, +, unii-45mz1t3tbv, r-methyl lactate
PubChem CID 637514
ChEBI CHEBI:74611
SMILES COC(=O)C(C)O

Specifications

Color Colorless
Density 1.0900g/mL
Boiling Point 144.0°C to 145.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4120 to 1.4140
Quantity 25 g
UN Number 3272
Specific Gravity 1.09
Specific Rotation Condition + 8.50 (20.00°C c=neat)
Specific Rotation + 8.50
Solubility Information Solubility in water: miscible,hydrolyses. Other solubilities: soluble in alcohol and ether,miscible with most common organic solvents
IUPAC Name methyl (2R)-2-hydroxypropanoate
Formula Weight 104.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl (R)-(+)-lactate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl (R)-(+)-lactate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.