missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyrrole-3-carboxylate, 97%

Product Code. 15490658
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490658 5 g 5g
15400668 250 mg 250mg
15480658 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490658 Supplier Thermo Scientific Alfa Aesar Supplier No. H54340.06
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2703-17-5 | C6H7NO2 | 125.127 g/mol

Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2703-17-5
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00967074
InChI Key WLBNVSIQCFHAQB-UHFFFAOYSA-N
Synonym methyl pyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylic acid, methyl ester, methyl 3-pyrrolecarboxylate, 3-methoxycarbonyl-1h-pyrrole, pubchem22507, acmc-1cmqn, methylpyrrole-3-carboxylate, 4,5-methoxy-carbonylpyrrole, methyl pyrrole 3-carboxylate
PubChem CID 12361759
IUPAC Name methyl 1H-pyrrole-3-carboxylate
SMILES COC(=O)C1=CNC=C1

Specifications

Melting Point 85°C to 89°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material Methyl pyrrole-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.