missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyrazine-2-carboxylate, 98+%, Thermo Scientific™

Catalog Number 11459892
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
6164-79-0
C6H6N2O2
138.126
MFCD00014611
TWIIRMSFZNYMQE-UHFFFAOYSA-N
methyl 2-pyrazinecarboxylate, methyl pyrazinoate, pyrazinoic acid methyl ester, methylpyrazine-2-carboxylate, pyrazinecarboxylic acid, methyl ester, methyl pyrazinecarboxylate, 2-pyrazinecarboxylic acid, methyl ester, pyrazine-2-carboxylic acid, methyl ester, 2-methylpyrazine carboxylate, pyrazine-2-carboxylic acid methyl ester
72662
methyl pyrazine-2-carboxylate
COC(=O)C1=NC=CN=C1
This item is not returnable. View return policy
6164-79-0
C6H6N2O2
138.126
MFCD00014611
TWIIRMSFZNYMQE-UHFFFAOYSA-N
methyl 2-pyrazinecarboxylate, methyl pyrazinoate, pyrazinoic acid methyl ester, methylpyrazine-2-carboxylate, pyrazinecarboxylic acid, methyl ester, methyl pyrazinecarboxylate, 2-pyrazinecarboxylic acid, methyl ester, pyrazine-2-carboxylic acid, methyl ester, 2-methylpyrazine carboxylate, pyrazine-2-carboxylic acid methyl ester
72662
methyl pyrazine-2-carboxylate
COC(=O)C1=NC=CN=C1
Melting Point 56°C to 60°C
Flash Point >100°C (212°F)
Quantity 5 g
Beilstein 118345
Merck Index 14,7959
Formula Weight 138.13
Percent Purity ≥98%
Chemical Name or Material Methyl pyrazine-2-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'