Learn More
Thermo Scientific Chemicals Methyl Purple, 0.1% w/v aq. soln.
Transition interval: pH 4.8 (purple) to pH 5.4 (green)
Brand: Thermo Scientific Chemicals 042753.K2
43.34 GBP valid until 2024-04-30
Use promo code "21637" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 1340-02-9 Weight : 1.00000kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Methyl Purple | |
0.1% w/v aqueous solution | |
Methyl purple: 0.1%; Water: 99.9% | |
1 L |
1340-02-9,7732-18-5 | |
MFCD00070843 | |
23 hPa (17mm Hg) at 20°C | |
Liquid |
RUO – Research Use Only