missing translation for 'onlineSavingsMsg'
Learn More

Methyl Propionylacetate, TRC

Product Code. 30481819
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481819 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30481819 Supplier Toronto Research Chemicals Supplier No. TRCM32682025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Propionylacetate
CAS 30414-53-0
Molecular Formula C6H10O3
Quantity 25 g
Synonym 3-Oxopentanoic Acid Methyl Ester, 3-oxo-Valeric Acid Methyl Ester, 3-Oxopentanoic Acid Methyl Ester, 3-Oxovaleric Acid Methyl Ester, Methyl 3-Oxopentanoate, Methyl 3-oxovalerate, Methyl Propanoylacetate, Methyl Propionylacetate, Methyl beta-Ketovalerate
InChI Formula InChI=1S/C6H10O3/c1-3-5(7)4-6(8)9-2/h3-4H2,1-2H3
SMILES CCC(=O)CC(=O)OC
IUPAC Name methyl 3-oxopentanoate
Molecular Weight (g/mol) 130.14
Formula Weight 130.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.