missing translation for 'onlineSavingsMsg'
Learn More

Methyl Propiolate, TRC

Product Code. 30466675
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30466675 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30466675 Supplier Toronto Research Chemicals Supplier No. TRCM3271001G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Propiolate
CAS 922-67-8
Molecular Formula C4 H4 O2
Quantity 1 g
Synonym 2-Propynoic Acid Methyl Ester, Propiolic Acid Methyl Ester, (Carbomethoxy)acetylene, (Methoxycarbonyl)acetylene, Methyl 2-Propynoate, Methyl Acetylenecarboxylate, Methyl Acetylenemonocarboxylate, Methyl Ethynecarboxylate, Methyl Propargylate, Methyl Propynoate, NSC 154164, Propynoic Acid Methyl Ester
InChI Formula InChI=1S/C4H4O2/c1-3-4(5)6-2/h1H,2H3
SMILES COC(=O)C#C
IUPAC Name methyl prop-2-ynoate
Molecular Weight (g/mol) 84.07
Formula Weight 84.0211
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.