missing translation for 'onlineSavingsMsg'
Learn More

Methyl Potassium Sulfate, MP Biomedicals

Product Code. 11422586 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11422586 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11422586 Supplier MP Biomedicals Supplier No. 0521548180
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 562-54-9
Molecular Formula CH3KO4S
MDL Number MFCD00040472
Quantity 100 g
Synonym potassium methyl sulfate, methyl potassium sulfate, methylsulfuric acid potassium salt, unii-9b0jf05o1c, sulfuric acid, monomethyl ester, potassium salt, potassium methyl sulphate, potassium monomethylsulfate, sulfuric acid, monomethyl ester, potassium salt 1:1, 75-93-4 parent, potassium methylsulfate
InChI Key WBGWGHYJIFOATF-UHFFFAOYSA-M
SMILES [K+].COS([O-])(=O)=O
IUPAC Name potassium methyl sulfate
Molecular Weight (g/mol) 150.19
PubChem CID 2733094
Formula Weight 150.2
Chemical Name or Material Methyl Potassium Sulfate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.