missing translation for 'onlineSavingsMsg'
Learn More

Methyl picolinate, 99%

Product Code. 15480808
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480808 100 g 100g
15470808 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480808 Supplier Thermo Scientific Alfa Aesar Supplier No. H54530.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2459-07-6 | C7H7NO2 | 137.14 g/mol

Methyl picolinate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl picolinate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7-6-2459
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00038038
InChI Key NMMIHXMBOZYNET-UHFFFAOYSA-N
Synonym methyl picolinate, 2-picolinic acid methyl ester, 2-pyridinecarboxylic acid, methyl ester, methyl 2-pyridinecarboxylate, picolinic acid, methyl ester, 2-carbomethoxypyridine, pyridine-2-carboxylic acid methyl ester, 2-pyridinecarboxylic acid methyl ester, picolinic acid methyl ester, methylpicolinate
PubChem CID 75569
IUPAC Name methyl pyridine-2-carboxylate
SMILES COC(=O)C1=CC=CC=N1

Specifications

Melting Point 19°C
Density 1.137
Boiling Point 95°C (1 mmHg)
Flash Point 103°C (217°F)
Refractive Index 1.521
Quantity 100 g
Solubility Information Slightly soluble in water.
Formula Weight 137.14
Percent Purity 99%
Chemical Name or Material Methyl picolinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.