missing translation for 'onlineSavingsMsg'
Learn More

Methyl (phenylthio)acetate, 99%, Thermo Scientific™

Product Code. 11393286
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393286 25 g 25g
11383286 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11393286 Supplier Thermo Scientific Alfa Aesar Supplier No. L12649.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17277-58-6
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00039805
InChI Key MUNSXQQODXYRKI-UHFFFAOYSA-N
Synonym methyl phenylthio acetate, methyl 2-phenylthio acetate, methyl phenylsulfanyl acetate, phenylthio acetic acid methyl ester, methyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, methyl ester, methyl 2-phenylthioacetate, pubchem10851
PubChem CID 123375
IUPAC Name methyl 2-phenylsulfanylacetate
SMILES COC(=O)CSC1=CC=CC=C1

Specifications

Density 1.17
Boiling Point 104°C to 105°C (4 mmHg)
Flash Point >110°C (230°F)
Odor Stench
Refractive Index 1.559
Quantity 25 g
Beilstein 2047554
Formula Weight 182.24
Percent Purity 99%
Chemical Name or Material Methyl (phenylthio)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.