missing translation for 'onlineSavingsMsg'
Learn More

Methyl (phenylthio)acetate, 99%, Thermo Scientific™

Product Code. 11383286
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11383286

Brand: Thermo Scientific Alfa Aesar L12649.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17277-58-6
Molecular Formula C9H10O2S
Molecular Weight (g/mol) 182.237
MDL Number MFCD00039805
InChI Key MUNSXQQODXYRKI-UHFFFAOYSA-N
Synonym methyl phenylthio acetate, methyl 2-phenylthio acetate, methyl phenylsulfanyl acetate, phenylthio acetic acid methyl ester, methyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, methyl ester, methyl 2-phenylthioacetate, pubchem10851
PubChem CID 123375
IUPAC Name methyl 2-phenylsulfanylacetate
SMILES COC(=O)CSC1=CC=CC=C1

Specifications

Density 1.17
Boiling Point 104°C to 105°C (4 mmHg)
Flash Point >110°C (230°F)
Odor Stench
Refractive Index 1.559
Quantity 5 g
Beilstein 2047554
Formula Weight 182.24
Percent Purity 99%
Chemical Name or Material Methyl (phenylthio)acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.