missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenylsulfonylacetate, 98%, Thermo Scientific Chemicals

£23.60 - £77.00

Chemical Identifiers

CAS 34097-60-4
Molecular Formula C9H10O4S
Molecular Weight (g/mol) 214.235
MDL Number MFCD00007555
InChI Key NLEAIFBNKPYTGN-UHFFFAOYSA-N
PubChem CID 555619
IUPAC Name methyl 2-(benzenesulfonyl)acetate
SMILES COC(=O)CS(=O)(=O)C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11306646
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L03748.06
5 g
£23.60
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11316646
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L03748.14
25 g
£77.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Methyl phenylsulfonylacetate has been used in a key step during the synthesis of (-)-hybridalactone, marine eicosanoid isolated from the red alga Laurencia hybrida, in dehydrative alkylation of alcohols under modified Mitsunobu conditions followed by desulfonylation using magnesium, in three step synthesis of 2-(hydroxymethyl)indene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl phenylsulfonylacetate has been used in a key step during the synthesis of (-)-hybridalactone, marine eicosanoid isolated from the red alga Laurencia hybrida, in dehydrative alkylation of alcohols under modified Mitsunobu conditions followed by desulfonylation using magnesium, in three step synthesis of 2-(hydroxymethyl)indene.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

34097-60-4
214.235
NLEAIFBNKPYTGN-UHFFFAOYSA-N
methyl 2-(benzenesulfonyl)acetate
C9H10O4S
MFCD00007555
555619
COC(=O)CS(=O)(=O)C1=CC=CC=C1

Specifications

34097-60-4
1.24
>110°C (230°F)
1.535
2051463
Insoluble in water.
COC(=O)CS(=O)(=O)C1=CC=CC=C1
214.235
214.24
Methyl phenylsulfonylacetate
∼30°C
165°C (0.05 mmHg)
C9H10O4S
MFCD00007555
methyl phenylsulfonylacetate, methyl 2-phenylsulfonyl acetate, methyl phenylsulfonyl acetate, methylphenylsulfonylacetate, benzenesulfonyl-acetic acid methyl ester, acmc-1ad0h, methyl benzenesulfonylacetate, methyl2-phenylsulfonyl acetate, methyl-phenylsulfonyl acetate
NLEAIFBNKPYTGN-UHFFFAOYSA-N
methyl 2-(benzenesulfonyl)acetate
555619
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only