missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenyl sulfoxide, 98+%, Thermo Scientific Chemicals

Catalog Number p-166870
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1193-82-4
C7H8OS
140.21
MFCD00002088
JXTGICXCHWMCPM-UHFFFAOYSA-N
methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
14516
methylsulfinylbenzene
CS(=O)C1=CC=CC=C1
This item is not returnable. View return policy
1193-82-4
C7H8OS
140.21
MFCD00002088
JXTGICXCHWMCPM-UHFFFAOYSA-N
methyl phenyl sulfoxide, methylsulfinyl benzene, thioanisole s-oxide, phenyl methyl sulfoxide, sulfoxide, methyl phenyl, benzene, methylsulfinyl, methanesulfinylbenzene, methylphenylsulfoxide, methylsulphinyl benzene
14516
methylsulfinylbenzene
CS(=O)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 34°C to 36°C
Color White
Boiling Point 92°C to 94°C (0.3 mmHg)
Flash Point 86°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula C6H5SOCH3
Refractive Index 1.5775
Quantity 5 g
Solubility Information Solubility in water: insoluble (20°C)
Formula Weight 140.21
Percent Purity 98+%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Methyl phenyl sulfoxide, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'