missing translation for 'onlineSavingsMsg'
Learn More

Methyl phenyl sulfone, 98+%

Product Code. 11409253
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409253 10 g 10g
11419253 50 g 50g
11429253 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409253 Supplier Thermo Scientific Alfa Aesar Supplier No. A11727.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3112-85-4 | C7H8O2S | 156.199 g/mol

Methyl phenyl sulfone, a versatile building block for the construction of chemicals with diverse functionalities

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl phenyl sulfone, a versatile building block for the construction of chemicals with diverse functionalities

Solubility
Soluble in methanol, ethanol, benzene, and chloroform. Partly miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3112-85-4
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.199
MDL Number MFCD00014741
InChI Key JCDWETOKTFWTHA-UHFFFAOYSA-N
Synonym methyl phenyl sulfone, methylsulfonyl benzene, phenyl methyl sulfone, benzene, methylsulfonyl, sulfone, methyl phenyl, phenylsulfonyl methane, methylphenylsulfone, methylsulphonyl benzene, methanesulfonylbenzene, methyl phenyl sulphone
PubChem CID 18369
IUPAC Name methylsulfonylbenzene
SMILES CS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 85°C to 88°C
Quantity 10 g
Beilstein 1906914
Solubility Information Soluble in methanol,ethanol,benzene,and chloroform. Partly miscible in water.
Formula Weight 156.2
Percent Purity ≥98%
Chemical Name or Material Methyl phenyl sulfone
compliance-icons
Product Identifier
  • Methyl phenyl sulphone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: (Methylsulphonyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.