missing translation for 'onlineSavingsMsg'
Learn More

Methyl Palmitate, TRC

Product Code. 30011936
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011936 10 g 10g
30004838 1 g 1g
30010243 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011936 Supplier Toronto Research Chemicals Supplier No. TRCM32558010G
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-39-0
Molecular Formula C17 H34 O2
Molecular Weight (g/mol) 270.45
Synonym Hexadecanoic Acid Methyl Ester, Hexadecanoic acid, methyl ester, Palmitic acid, methyl ester (6CI,8CI), Edenor ME C16-98, Edenor ME-C 16-80MY, Emery 2216, Metholene 2216, Methyl hexadecanoate, Methyl n-hexadecanoate, Methyl palmitate, NSC 4197, Pastell M 16, Radia 7120, Uniphat A60, n-Hexadecanoic acid methyl ester
IUPAC Name methyl hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C17H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h3-16H2,1-2H3
Formula Weight 270.2559
Chemical Name or Material Methyl Palmitate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.