missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl palmitate, 95%, Thermo Scientific Chemicals
£16.70 - £51.60
Chemical Identifiers
CAS | 112-39-0 |
---|---|
Molecular Formula | C17H34O2 |
Molecular Weight (g/mol) | 270.45 |
MDL Number | MFCD00008994 |
InChI Key | FLIACVVOZYBSBS-UHFFFAOYSA-N |
Synonym | methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i |
PubChem CID | 8181 |
ChEBI | CHEBI:69187 |
IUPAC Name | methyl hexadecanoate |
SMILES | CCCCCCCCCCCCCCCC(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10635871
|
Thermo Scientific Acros
414681000 |
100 g | Glass bottle |
£16.70
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10676051
|
Thermo Scientific Acros
414685000 |
500 g | Glass bottle |
£51.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-39-0 | |
270.45 | |
FLIACVVOZYBSBS-UHFFFAOYSA-N | |
8181 | |
methyl hexadecanoate |
C17H34O2 | |
MFCD00008994 | |
methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i | |
CHEBI:69187 | |
CCCCCCCCCCCCCCCC(=O)OC |
Specifications
112-39-0 | |
Colorless | |
338.0°C | |
Authentic | |
C17H34O2 | |
MFCD00008994 | |
Solubility in water: insoluble. Other solubilities: solubel in alcohol,ether | |
CCCCCCCCCCCCCCCC(=O)OC | |
270.45 | |
CHEBI:69187 | |
95% | |
Methyl palmitate, 95% |
28.0°C | |
0.85g/mL | |
>110°C | |
94% min. (GC) | |
CH3(CH2)14CO2CH3 | |
methyl palmitate, palmitic acid methyl ester, hexadecanoic acid, methyl ester, palmitic acid, methyl ester, uniphat a60, metholene 2216, methyl n-hexadecanoate, hexadecanoic acid methyl ester, n-hexadecanoic acid methyl ester, unii-dpy8vcm98i | |
FLIACVVOZYBSBS-UHFFFAOYSA-N | |
methyl hexadecanoate | |
8181 | |
270.45 | |
Liquid or Low Melting Solid |
Safety and Handling
missing translation for 'einecsNumber' : 203-966-3
missing translation for 'tsca' : TSCA
RUO – Research Use Only