missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-tolyl sulfone, 99%, Thermo Scientific Chemicals

Product Code. 11404543
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3185-99-7
Molecular Formula C8H10O2S
Molecular Weight (g/mol) 170.23
MDL Number MFCD00014742
InChI Key YYDNBUBMBZRNQQ-UHFFFAOYSA-N
Synonym methyl p-tolyl sulfone, 1-methyl-4-methylsulfonyl benzene, 4-methylsulfonyl toluene, benzene, 1-methyl-4-methylsulfonyl, p-tolyl methyl sulfone, sulfone, methyl p-tolyl, 4-methylsulphonyl toluene, 1-methanesulfonyl-4-methyl-benzene, unii-tg77sm513a, p-methylsulfonyltoluene
PubChem CID 18521
IUPAC Name 1-methyl-4-methylsulfonylbenzene
SMILES CC1=CC=C(C=C1)S(C)(=O)=O

Specifications

Melting Point 86°C to 89°C
Boiling Point 140°C (3 mmHg)
Quantity 5 g
Beilstein 607296
Formula Weight 170.23
Percent Purity 99%
Chemical Name or Material Methyl p-tolyl sulfone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'