missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-Toluenesulfonate, TRC

Product Code. 30477157
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477157 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30477157 Supplier Toronto Research Chemicals Supplier No. TRCM3306655G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Toluene-4-sulphonate
CAS 80-48-8
Purity Grade Notes HPLC
Molecular Formula C8 H10 O3 S
Quantity 5 g
Synonym Methyl Toluene-4-sulphonate, Benzenesulfonic acid, 4-methyl-, methyl ester (9CI, ACI), p-Toluenesulfonic acid, methyl ester (6CI, 7CI, 8CI), Methyl 4-methylbenzenesulfonate, Methyl p-methylbenzenesulfonate, Methyl p-toluenesulfonate, Methyl p-tosylate, Methyl toluene-4-sulfonate, Methyl toluenesulfonate, Methyl tosylate, NSC 406335, p-Methylbenzenesulfonic acid methyl ester, Toluene-4-sulfonic acid methyl ester
InChI Formula InChI=1S/C8H10O3S/c1-7-3-5-8(6-4-7)12(9,10)11-2/h3-6H,1-2H3
SMILES COS(=O)(=O)c1ccc(C)cc1
IUPAC Name methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 186.23
Formula Weight 186.0351
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.