missing translation for 'onlineSavingsMsg'
Learn More

Methyl p-toluenesulfonate, 98%

Product Code. 11474233
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474233 1000 g 1000g
11484233 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474233 Supplier Thermo Scientific Alfa Aesar Supplier No. A10881.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-48-8 | C8H10O3S | 186.225 g/mol

Methyl p-toluenesulfonate is used as a methylating agent in organic synthesis. It acts as a catalyst for alkyd resins. It is also employed in selective 1-substitution reaction of tetrazole. Further, it is used in the preparation of dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl p-toluenesulfonate is used as a methylating agent in organic synthesis. It acts as a catalyst for alkyd resins. It is also employed in selective 1-substitution reaction of tetrazole. Further, it is used in the preparation of dyes.

Solubility
Soluble in ether, alcohol, benzene and chloroform. Slightly soluble in petroleum ether. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-48-8
Molecular Formula C8H10O3S
Molecular Weight (g/mol) 186.225
MDL Number MFCD00008417
InChI Key VUQUOGPMUUJORT-UHFFFAOYSA-N
Synonym methyl p-toluenesulfonate, methyl tosylate, methyl p-tosylate, methyl toluene-4-sulfonate, methyl-p-toluenesulfonate, p-toluenesulfonic acid, methyl ester, benzenesulfonic acid, 4-methyl-, methyl ester, methyl toluene-4-sulphonate, p-toluenesulfonic acid methyl ester, p-methylbenzenesulfonate methyl ester
PubChem CID 6645
IUPAC Name methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC

Specifications

Melting Point 24°C to 28°C
Density 1.234
Boiling Point 140°C (20 mmHg)
Flash Point 152°C (305°F)
Refractive Index 1.5185
Quantity 1000 g
UN Number UN3261
Beilstein 609209
Solubility Information Soluble in ether,alcohol,benzene and chloroform. Slightly soluble in petroleum ether. Insoluble in water.
Formula Weight 186.23
Percent Purity 98%
Chemical Name or Material Methyl p-toluenesulfonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl p-toluenesulfonate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Methyl p-toluenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.